4-Chlorbenzylisothiocyanat ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1-Chloro-4-(isothiocyanatomethyl)benzene
L09579.03
L09579.06
L09579.03EA
41.7
EUR
L09579.03
L09579.06
4-Chlorbenzylisothiocyanat ≥97%
4-Chlorbenzylisothiocyanat
Purity: 97%
CAS No.: 3694-45-9
Molecular Formula: C8H6ClNS
Formula Weight: 183.66
Storage Temperature: 2°C to 8°C
Refractive Index: 1.6145
Odour: Irritating
Boiling Point: 169-170°/25mm
Beilstein Reference: 2085917
UN No.: 2922
MDL No.: MFCD00041116
Formel:
C₈H₆ClNS Molecular Weight: 183,66 g/mol Siedepunkt: 169…170 °C (25 mmHg) Dichte: 1,270 |
MDL:
MFCD00041116 CAS-Nummer: 3694-45-9 ADR: 8,III |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)