Thiazol 99
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L09970.03
L09970.06
L09970.14
L09970.03EA
35.7
EUR
L09970.03
L09970.06
L09970.14
Thiazol 99
Thiazole
Purity: 99%
CAS No.: 288-47-1
Molecular Formula: C3H3NS
Formula Weight: 85.13
Flash Point: 22°(71°F)
Refractive Index: 1.5390
Odour: Unpleasant
Melting Point: -33°
Boiling Point: 117-118°
Beilstein Reference: 103852
Merck Reference: 14,9307
UN No.: 1993
MDL No.: MFCD00005315
Formel:
C₃H₃NS Molecular Weight: 85.13 g/mol Siedepunkt: 117…118 °C Schmelzpunkt: –33 °C Dichte: 1.200 Flammpunkt: 22 °C (71 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005315 CAS-Nummer: 288-47-1 EINECS: 206-021-3 ADR: 3 II Merck Index: 12,09442 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...