1-(3-Hydroxyphenyl)-2-thioharnstoff ≥97%

Lieferant: Thermo Scientific
Achtung

L09975.09 L09975.04
L09975.09EA 238 EUR
L09975.09 L09975.04
1-(3-Hydroxyphenyl)-2-thioharnstoff ≥97%
1-(3-Hydroxyphenyl)-2-thioharnstoff

Purity: 97% CAS No.: 3394-05-6 Molecular Formula: C7H8N2OS Formula Weight: 168.22 Odour: Odourless Melting Point: 175-180° Beilstein Reference: 2090372 MDL No.: MFCD00022167

Formel: (C₆H₄(OH))NC(S)NH₂
Molecular Weight: 168,22 g/mol
Schmelzpunkt: 175…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00022167
CAS-Nummer: 3394-05-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR