2,6-Dichlorbenzothiazol ≥97%
Lieferant: Thermo Scientific
L10256.03
L10256.06
L10256.03EA
24.8
EUR
L10256.03
L10256.06
2,6-Dichlorbenzothiazol ≥97%
2,6-Dichlorobenzothiazole
Purity: 97%
CAS No.: 3622-23-9
Molecular Formula: C7H3Cl2NS
Formula Weight: 204.08
Melting Point: 94-98°
Beilstein Reference: 127749
Formel:
C₇H₃Cl₂NS Molecular Weight: 204,08 g/mol Schmelzpunkt: 94…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00044101 CAS-Nummer: 3622-23-9 EINECS: 222-819-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...