4-Chlor-2-methylphenylisocyanat ≥98%

Lieferant: Thermo Scientific
Gefahr

L10470.03 L10470.06
L10470.03EA 22.8 EUR
L10470.03 L10470.06
4-Chlor-2-methylphenylisocyanat ≥98%
4-Chlor-2-methylphenylisocyanat

Purity: 98% CAS No.: 37408-18-7 Molecular Formula: C8H6ClNO Formula Weight: 167.60 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 36-40° Boiling Point: 103°/13mm Beilstein Reference: 2803097 UN No.: 2811 MDL No.: MFCD00019914

Formel: C₈H₆ClNO
Molecular Weight: 167,59 g/mol
Siedepunkt: 103 °C (13 mmHg)
Schmelzpunkt: 36…40 °C
Flammpunkt: >110 °C (230 °F)
MDL: MFCD00019914
CAS-Nummer: 37408-18-7
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR