3-Chlor-2-methylphenylisocyanat ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Isocyansäure(3-chlor-2-methylphenyl)ester
L10802.03
L10802.06
L10802.03EA
25.6
EUR
L10802.03
L10802.06
3-Chlor-2-methylphenylisocyanat ≥99%
3-Chlor-2-methylphenylisocyanat
Purity: 99%
CAS No.: 40397-90-8
Molecular Formula: C8H6ClNO
Formula Weight: 167.60
Storage Temperature: 2°C to 8°C
Flash Point:
Formel:
C₈H₆ClNO Molecular Weight: 167.59 g/mol Siedepunkt: 94…95 °C (9 mmHg) Dichte: 1.247 Flammpunkt: >110 °C (230 °F) |
MDL:
MFCD00037036 CAS-Nummer: 40397-90-8 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...