4-Isopropylphenylisocyanat ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
L11217.09
L11217.04
L11217.09EA
152
EUR
L11217.09
L11217.04
4-Isopropylphenylisocyanat ≥98%
4-Isopropylphenylisocyanat
Purity: 98%
CAS No.: 31027-31-3
Molecular Formula: C10H11NO
Formula Weight: 161.21
Storage Temperature: 2°C to 8°C
Flash Point: 90°(194°F)
Refractive Index: 1.5205
Boiling Point: 84-85°/4mm
Beilstein Reference: 2936403
UN No.: 2206
MDL No.: MFCD00013880
Formel:
C₁₀H₁₁NO Molecular Weight: 161,2 g/mol Siedepunkt: 84…85 °C (4 mmHg) Dichte: 1,010 Flammpunkt: 90 °C (194 °F) |
MDL:
MFCD00013880 CAS-Nummer: 31027-31-3 EINECS: 250-439-9 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...