2-Amino-4-methoxybenzothiazol 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Methoxybenzothiazol-2-ylamine

L11358.03 L11358.06 L11358.14
L11358.03EA 40.4 EUR
L11358.03 L11358.06 L11358.14
2-Amino-4-methoxybenzothiazol 97%
2-Amino-4-methoxybenzothiazole

Purity: 97% CAS No.: 5464-79-9 Molecular Formula: C8H8N2OS Formula Weight: 180.23 Melting Point: 153-159° Beilstein Reference: 141363

Formel: C₈H₈N₂OS
Molecular Weight: 180.23 g/mol
Siedepunkt: 335
Schmelzpunkt: 153…159 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005792
CAS-Nummer: 5464-79-9
EINECS: 226-763-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR