1-Chlor-3,3-dimethylbutan ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
L11753.09
L11753.18
L11753.09EA
77.2
EUR
L11753.09
L11753.18
1-Chlor-3,3-dimethylbutan ≥97%
1-Chloro-3,3-dimethylbutane
Purity: 97%
CAS No.: 2855-08-5
Molecular Formula: C6H13Cl
Formula Weight: 120.62
Flash Point: 7°(44°F)
Refractive Index: 1.4170
Boiling Point: 116-118°
Beilstein Reference: 1731465
UN No.: 1993
MDL No.: MFCD00018980
Formel:
C₆H₁₃Cl Siedepunkt: 116…118 °C Dichte: 0,867 Flammpunkt: 7 °C (44 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00018980 CAS-Nummer: 2855-08-5 EINECS: 220-665-2 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...