trans-5-Decen-1-ol 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: trans-1-Hydroxy-5-decen, trans-Dec-5-en-1-ol

L11768.03 L11768.06
L11768.03EA 68.3 EUR
L11768.03 L11768.06
trans-5-Decen-1-ol 96%
trans-5-Decen-1-ol

Purity: 96% CAS No.: 56578-18-8 Molecular Formula: C10H20O Formula Weight: 156.27 Flash Point: 62°(143°F) Refractive Index: 1.4520 Boiling Point: 113-118°/11mm Beilstein Reference: 1840630 MDL No.: MFCD00056187

Formel: C₁₀H₂₀O
Molecular Weight: 156,27 g/mol
Siedepunkt: 113…118 °C (11 mmHg)
Flammpunkt: 62 °C (143 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00056187
CAS-Nummer: 56578-18-8
EINECS: 260-267-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR