2-Chlor-4-fluorbenzylamin 97%

Lieferant: Thermo Fisher Scientific
Gefahr

L12957.03 L12957.06
L12957.03EA 76.4 EUR
L12957.03 L12957.06
2-Chlor-4-fluorbenzylamin 97%
2-Chlor-4-fluorbenzylamin

Purity: 97% CAS No.: 15205-11-5 Molecular Formula: C7H7ClFN Formula Weight: 159.59 Flash Point: 110°(230°F) Refractive Index: 1.5350 Boiling Point: 94-95°/15mm Beilstein Reference: 2936421 UN No.: 2735 MDL No.: MFCD00042532

Formel: C₇H₇ClFN
Siedepunkt: 94…95 °C (15 mmHg)
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00042532
CAS-Nummer: 15205-11-5
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR