2,2'-(Phenylimino)diethanol 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
N-Phenyldiethanolamin, N-Phenyl-2,2'-iminodiethanol
L13931.22
L13931.36
L13931.0E
L13931.22EA
19.6
EUR
L13931.22
L13931.36
L13931.0E
2,2'-(Phenylimino)diethanol 97%
2,2'-Phenyliminodiethanol
Purity: 97%
CAS No.: 120-07-0
Molecular Formula: C10H15NO2
Formula Weight: 181.23
Flash Point: 178°(352°F)
Odour: Amine-like
pH: 8.2
Melting Point: 55-59°
Boiling Point: 270°
Beilstein Reference: 2096832
MDL No.: MFCD00002845
Formel:
(C₆H₅)N(CH₂CH₂OH)₂ Molecular Weight: 181,23 g/mol Siedepunkt: 270 °C Schmelzpunkt: 55…59 °C Dichte: 1,1 Flammpunkt: 178 °C (352 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002845 CAS-Nummer: 120-07-0 EINECS: 204-368-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...