3',5'-Dimethylacetophenon ≥97%
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
1-(3,5-Dimethylphenyl)ethan-1-on
L15480.03
L15480.06
L15480.03EA
49.4
EUR
L15480.03
L15480.06
3',5'-Dimethylacetophenon ≥97%
3',5'-Dimethylacetophenon
Purity: 97%
CAS No.: 5379-16-8
Molecular Formula: C10H12O
Formula Weight: 148.21
Flash Point: 93°(199°F)
Refractive Index: 1.5280
Odour: Characteristic
Melting Point: ca 20°
Boiling Point: 238°
Beilstein Reference: 2240987
MDL No.: MFCD01075693
Formel:
C₁₀H₁₂O Molecular Weight: 148.2 g/mol Siedepunkt: 238 °C Schmelzpunkt: ∼ 20 °C Dichte: 0.988 Flammpunkt: 93 °C (199 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075693 CAS-Nummer: 5379-16-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...