3-(Hydroxymethyl)indol ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: 3-Indolcarbinol, Indol-3-carbinol, (Indol-3-yl)methanol, 3-Indolmethanol, (3-Indolyl)methanol, Indol-3-methanol

L15762.03 L15762.06
L15762.03EA 24.5 EUR
L15762.03 L15762.06
3-(Hydroxymethyl)indol ≥97%
3-(Hydroxymethyl)indole

Purity: 97% CAS No.: 700-06-1 Molecular Formula: C9H9NO Formula Weight: 147.18 Melting Point: 96-99° Beilstein Reference: 121323

Formel: C₉H₉NO
Molecular Weight: 147.18 g/mol
Schmelzpunkt: 96…99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005632
CAS-Nummer: 700-06-1
EINECS: 211-836-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR