5-Methylthiazol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
L15879.06
L15879.14
L15879.06EA
183
EUR
L15879.06
L15879.14
5-Methylthiazol ≥97%
5-Methylthiazole
Purity: 97%
CAS No.: 3581-89-3
Molecular Formula: C4H5NS
Formula Weight: 99.16
Flash Point: 42°(107°F)
Refractive Index: 1.5280
Odour: Stench
Boiling Point: 141-142°
Beilstein Reference: 104674
UN No.: 1993
MDL No.: MFCD00005335
Formel:
C₄H₅NS Molecular Weight: 99,16 g/mol Siedepunkt: 141…142 °C Schmelzpunkt: –40,4 °C Dichte: 1,120 Flammpunkt: 42 °C (107 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005335 CAS-Nummer: 3581-89-3 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...