3-(2-Aminoethyl)pyridin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-(3-Pyridyl)ethylamin

L16311.03 L16311.06
L16311.03EA 109 EUR
L16311.03 L16311.06
3-(2-Aminoethyl)pyridin 98%
3-(2-Aminoethyl)pyridine

Purity: 98% CAS No.: 20173-24-4 Molecular Formula: C7H10N2 Formula Weight: 122.17 Refractive Index: 1.5518 Odour: Amine-like Boiling Point: 119-120°/19mm Beilstein Reference: 111288 UN No.: 2735 MDL No.: MFCD00191604

Formel: C₇H₁₀N₂
Molecular Weight: 122.17 g/mol
Siedepunkt: 119…120 °C (19 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00191604
CAS-Nummer: 20173-24-4
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR