ω-Chlor-ω,ω-difluoracetophenon 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Chlor-2,2-difluoracetophenon, α-Chlor-α,α-difluoracetophenon

L16963.06 AB180285.0005
L16963.06EA 99.6 EUR
L16963.06 ABCRAB180285.0005
ω-Chlor-ω,ω-difluoracetophenon 96%
ω-Chlor-ω,ω-difluoracetophenon

Purity: 96% CAS No.: 384-67-8 Molecular Formula: C8H5ClF2O Formula Weight: 190.58 Flash Point: 72°(161°F) Refractive Index: 1.4970 Boiling Point: 95-97°/37mm Beilstein Reference: 2251448 MDL No.: MFCD00042123

Formel: (C₆H₅)C(O)CClF₂
Molecular Weight: 190,58 g/mol
Siedepunkt: 95…97 °C (37 mmHg)
Dichte: 1,293
Flammpunkt: 72 °C (161 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00042123
CAS-Nummer: 384-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR