7-Indolcarbaldehyd ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Indol-7-carboxaldehyd

L17506.03 L17506.MD
L17506.03EA 89.4 EUR
L17506.03 L17506.MD
7-Indolcarbaldehyd ≥98%
7-Indolcarbaldehyd

Purity: 98% CAS No.: 1074-88-0 Molecular Formula: C9H7NO Formula Weight: 145.16 Melting Point: 86-89° Beilstein Reference: 1524960

Formel: C₉H₇NO
Molecular Weight: 145,16 g/mol
Schmelzpunkt: 86…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318152
CAS-Nummer: 1074-88-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR