3-(3-Bromphenyl)propionsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L18080.03
L18080.06
L18080.03EA
39
EUR
L18080.03
L18080.06
3-(3-Bromphenyl)propionsäure ≥97%
3-(3-Bromphenyl)propionsäure
Purity: 97%
CAS No.: 42287-90-1
Molecular Formula: C9H9BrO2
Formula Weight: 229.07
Melting Point: 63-65°
Beilstein Reference: 2362753
Formel:
C₉H₉BrO₂ Schmelzpunkt: 63…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01310792 CAS-Nummer: 42287-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...