Oxazol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L18341.03
L18341.06
AB180490.0005
L18341.03EA
81.1
EUR
L18341.03
L18341.06
ABCRAB180490.0005
Oxazol ≥98%
Oxazole
Purity: 98+%
CAS No.: 288-42-6
Molecular Formula: C3H3NO
Formula Weight: 69.07
Storage Temperature: 2°C to 8°C
Flash Point: 19°(66°F)
Refractive Index: 1.4270
Melting Point: -86°
Boiling Point: 69-70°
Beilstein Reference: 103851
UN No.: 2924
MDL No.: MFCD00009751
Formel:
C₃H₃NO Siedepunkt: 69…70 °C Schmelzpunkt: –86 °C Dichte: 1,050 Flammpunkt: 19 °C (66 °F) |
MDL:
MFCD00009751 CAS-Nummer: 288-42-6 EINECS: 206-020-8 UN: 2924 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...