5-Brom-2-chlorpyrimidin ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
L18360.06
L18360.14
L18360.06EA
114
EUR
L18360.06
L18360.14
5-Brom-2-chlorpyrimidin ≥98%
5-Bromo-2-chloropyrimidine
Purity: 98+%
CAS No.: 32779-36-5
Molecular Formula: C4H2BrClN2
Formula Weight: 193.43
Melting Point: 77-80°
Boiling Point: 95°/15mm
MDL No.: MFCD00483232
Formel:
C₄H₂BrClN₂ Molecular Weight: 193,43 g/mol Siedepunkt: 95 °C (15 mmHg) Schmelzpunkt: 77…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00483232 CAS-Nummer: 32779-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...