6-Aminonicotinonitril 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Amino-5-pyridincarbonsäurenitril, 2-Amino-5-cyanpyridin, 6-Aminonicotinsäurenitril, 2-Amino-5-pyridincarbonitril
L19282.06
L19282.14
L19282.06EA
33.5
EUR
L19282.06
L19282.14
6-Aminonicotinonitril 98%
6-Aminonicotinonitril
Purity: 98%
CAS No.: 4214-73-7
Molecular Formula: C6H5N3
Formula Weight: 119.13
Melting Point: 159-164°
Boiling Point: 240-250°/15mm
UN No.: 3439
MDL No.: MFCD00128902
Formel:
C₆H₅N₃ Molecular Weight: 119,13 g/mol Siedepunkt: 240…250 °C (15 mmHg) Schmelzpunkt: 159…164 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128902 CAS-Nummer: 4214-73-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...