1-(4-Pyridyl)piperazin ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4-(1-Piperazinyl)pyridin

L19396.06 L19396.03 L19396.14
L19396.06EA 125 EUR
L19396.06 L19396.03 L19396.14
1-(4-Pyridyl)piperazin ≥97%
1-(4-Pyridyl)piperazin

Purity: 97% CAS No.: 1008-91-9 Molecular Formula: C9H13N3 Formula Weight: 163.22 Melting Point: 136-141° Boiling Point: 195-200°/0.3mm UN No.: 3259 MDL No.: MFCD00040745

Formel: C₉H₁₃N₃
Molecular Weight: 163,22 g/mol
Siedepunkt: 195…200 °C (0,3 mmHg)
Schmelzpunkt: 136…141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040745
CAS-Nummer: 1008-91-9
EINECS: 213-764-7
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR