2-Amino-6-(trifluormethyl)pyridin ≥98%

Lieferant: Thermo Scientific
Gefahr

L19443.MC L19443.ME
L19443.MCEA 47.6 EUR
L19443.MC L19443.ME
2-Amino-6-(trifluormethyl)pyridin ≥98%
2-Amino-6-(trifluoromethyl)pyridine

Purity: 98+% CAS No.: 34486-24-3 Molecular Formula: C6H5F3N2 Formula Weight: 162.11 Melting Point: 86-89° Beilstein Reference: 473259 UN No.: 2811 MDL No.: MFCD03093868

Formel: C₆H₅F₃N₂
Molecular Weight: 162,11 g/mol
Siedepunkt: 200 °C (1013 hPa)
Schmelzpunkt: 86…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03093868
CAS-Nummer: 34486-24-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR