2-Aminoisonicotinonitril 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Aminopyridine-4-carbonitrile, 2-Amino-4-cyanopyridine

L19841.03 L19841.06 L19841.14
L19841.03EA 42.3 EUR
L19841.03 L19841.06 L19841.14
2-Aminoisonicotinonitril 97%
2-Aminoisonicotinonitril

Purity: 97% CAS No.: 42182-27-4 Molecular Formula: C6H5N3 Formula Weight: 119.13 Melting Point: 148-152° Beilstein Reference: 386393 UN No.: 3439 MDL No.: MFCD03791310

Formel: C₆H₅N₃
Molecular Weight: 119.13 g/mol
Schmelzpunkt: 148…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03791310
CAS-Nummer: 42182-27-4
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR