4-Benzylanilin ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Aminodiphenylmethane
L19873.03
L19873.06
L19873.03EA
53.6
EUR
L19873.03
L19873.06
4-Benzylanilin ≥98%
4-Benzylanilin
Purity: 98%
CAS No.: 1135-12-2
Molecular Formula: C13H13N
Formula Weight: 183.25
Melting Point: 35-38°
Boiling Point: 142-144°/1mm
MDL No.: MFCD00047861
Formel:
C₁₃H₁₃N Molecular Weight: 183,25 g/mol Siedepunkt: 142…144 °C (1 mmHg) Schmelzpunkt: 35…38 °C Dichte: 1,07 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047861 CAS-Nummer: 1135-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...