D(-)-allo-Isoleucin

Lieferant: BACHEM BIOCHEMICA

Synonyme: (2R,3S)-(-)-α-Amino-β-methylvaleriansäure, (2R,3S)-(-)-allo-Isoleucin, (2R,3S)-(-)-2-Amino-3-methylpentansäure, H-D-allo-Ile-OH

4015231.0250 4015231.1000 4015231.5000
BACE4015231.0250EA 0 EUR
BACE4015231.0250 BACE4015231.1000 BACE4015231.5000
D(-)-allo-Isoleucin
D(-)-allo-Isoleucin
Formel: C₆H₁₃NO₂
Molecular Weight: 131.17 g/mol
MDL: MFCD00066445
CAS-Nummer: 1509-35-9
EINECS: 216-143-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR