D(-)-Isoleucin

Lieferant: BACHEM BIOCHEMICA

Synonyme: (2R,3R)-(-)-α-Amino-β-methylvaleriansäure, (2R,3R)-(-)-2-Amino-3-methylpentansäure, (2R,3R)-(-)-Isoleucin, H-D-Ile-OH

4006515.0001 4006515.0005
BACE4006515.0001EA 0 EUR
BACE4006515.0001 BACE4006515.0005
D(-)-Isoleucin
D(-)-Isoleucin
Formel: H₃CCH₂CH(CH₃)CH(NH₂)COOH
Molecular Weight: 131.17 g/mol
Siedepunkt: 235
Schmelzpunkt: 98
Dichte: 1.224 g/cm³ (20 °C)
MDL: MFCD00064221
CAS-Nummer: 319-78-8
EINECS: 206-269-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR