3-Amino-1H-indazol

Lieferant: BIONET RESEARCH

Synonyme: 3-Amino-1H-indazol, Indazol-3-ylamin, 1H-Indazol-3-ylamin, 1H-Indazol-3-amin, Indazol-3-amin

10P-212.500 10P-212.1 10P-212.5 10P-212.10
BIOT10P-212.500EA 0 EUR
BIOT10P-212.500 BIOT10P-212.1 BIOT10P-212.5 BIOT10P-212.10
3-Amino-1H-indazol
3-Amino-1H-indazole
Formel: C₇H₇N₃
Molecular Weight: 133,15 g/mol
MDL: MFCD00182045
CAS-Nummer: 874-05-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR