6-Chlornicotinonitril

Lieferant: BIONET RESEARCH

Synonyme: 2-Chlor-5-pyridincarbonitril, 6-Chlornicotinsäurenitril, 2-Chlor-5-cyanpyridin, 2-Chlor-5-pyridincarbonsäurenitril

11X-0845.500 11X-0845.1 11X-0845.5 11X-0845.10
BIOT11X-0845.500EA 0 EUR
BIOT11X-0845.500 BIOT11X-0845.1 BIOT11X-0845.5 BIOT11X-0845.10
6-Chlornicotinonitril
6-Chlornicotinonitril
Formel: C₆H₃ClN₂
Molecular Weight: 138,56 g/mol
MDL: MFCD00084941
CAS-Nummer: 33252-28-7
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR