2-Methyl-6-nitrobenzoxazol

Lieferant: BIONET RESEARCH

12T-0885.500 12T-0885.1 12T-0885.5 12T-0885.10
BIOT12T-0885.500EA 0 EUR
BIOT12T-0885.500 BIOT12T-0885.1 BIOT12T-0885.5 BIOT12T-0885.10
2-Methyl-6-nitrobenzoxazol
2-Methyl-6-nitrobenzoxazole
Formel: C₈H₆N₂O₃
Molecular Weight: 178.15 g/mol
MDL: MFCD00022848
CAS-Nummer: 5683-43-2
EINECS: 227-149-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR