2-Acetyl-3-chlorthiophen

Lieferant: BIONET RESEARCH

Synonyme: 1-(3-Chloro-2-thienyl)-1-ethanone

1J-931.1 1J-931.10 1J-931.5 1J-931.500
BIOT1J-931.1EA 0 EUR
BIOT1J-931.1 BIOT1J-931.10 BIOT1J-931.5 BIOT1J-931.500
2-Acetyl-3-chlorthiophen
2-Acetyl-3-chlorthiophen
Formel: C₆H₅ClOS
Molecular Weight: 160,62 g/mol
MDL: MFCD00794248
CAS-Nummer: 89581-82-8
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR