2-Methyl-5-nitrobenzoxazol

Lieferant: BIONET RESEARCH

1W-0929.500 1W-0929.1 1W-0929.5 1W-0929.10
BIOT1W-0929.500EA 0 EUR
BIOT1W-0929.500 BIOT1W-0929.1 BIOT1W-0929.5 BIOT1W-0929.10
2-Methyl-5-nitrobenzoxazol
2-Methyl-5-nitrobenzoxazole
Formel: C₈H₆N₂O₃
MDL: MFCD00466748
CAS-Nummer: 32046-51-8
EINECS: 250-904-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR