2-Chlorisonicotinonitril

Lieferant: BIONET RESEARCH

Synonyme: 2-Chlor-4-cyanpyridin, 2-Chlor-4-pyridincarbonitril, 2-Chlor-4-pyridincarbonsäurenitril

1Y-0715.1 1Y-0715.10 1Y-0715.5 1Y-0715.500
BIOT1Y-0715.1EA 0 EUR
BIOT1Y-0715.1 BIOT1Y-0715.10 BIOT1Y-0715.5 BIOT1Y-0715.500
2-Chlorisonicotinonitril
2-Chlorisonicotinonitril
Formel: C₆H₃ClN₂
Molecular Weight: 138,56 g/mol
MDL: MFCD00174318
CAS-Nummer: 33252-30-1
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR