1-(3-Bromphenyl)pyrrol
Lieferant: BIONET RESEARCH
Synonyme:
N-(3-Bromphenyl)pyrrol
2Y-0843.1
2Y-0843.10
2Y-0843.5
2Y-0843.500
BIOT2Y-0843.1EA
0
EUR
BIOT2Y-0843.1
BIOT2Y-0843.10
BIOT2Y-0843.5
BIOT2Y-0843.500
1-(3-Bromphenyl)pyrrol
1-(3-Bromophenyl)pyrrole
Formel:
C₁₀H₈BrN Molecular Weight: 222,08 g/mol |
MDL:
MFCD02665243 CAS-Nummer: 107302-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...