1-(2-Bromethoxy)-2,4-dichlorbenzol
Lieferant: BIONET RESEARCH
Synonyme:
beta-Bromo-2,4-dichlorophenetole
3F-947.500
3F-947.1
3F-947.5
3F-947.10
BIOT3F-947.500EA
0
EUR
BIOT3F-947.500
BIOT3F-947.1
BIOT3F-947.5
BIOT3F-947.10
1-(2-Bromethoxy)-2,4-dichlorbenzol
1-(2-Bromoethoxy)-2,4-dichlorobenzene
Formel:
C₈H₇BrCl₂O Molecular Weight: 269.95 g/mol |
MDL:
MFCD00043464 CAS-Nummer: 6954-77-4 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...