7-Amino-1H-indazol

Lieferant: BIONET RESEARCH

3P-091.500 3P-091.1 3P-091.5 3P-091.10
BIOT3P-091.500EA 0 EUR
BIOT3P-091.500 BIOT3P-091.1 BIOT3P-091.5 BIOT3P-091.10
7-Amino-1H-indazol
7-Amino-1H-indazole
Formel: C₇H₇N₃
Molecular Weight: 133,15 g/mol
MDL: MFCD00022790
CAS-Nummer: 21443-96-9
EINECS: 244-391-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR