3-(2-Hydroxyethyl)-5-methoxyindol
Lieferant: BIONET RESEARCH
Synonyme:
5-Methoxyindol-3-ethanol, 5-Methoxytryptophol, 2-(5-Methoxy-1H-indol-3-yl)-1-ethanol
3Y-5037.1
3Y-5037.5
3Y-5037.500
3Y-5037.10
BIOT3Y-5037.1EA
0
EUR
BIOT3Y-5037.1
BIOT3Y-5037.5
BIOT3Y-5037.500
BIOT3Y-5037.10
3-(2-Hydroxyethyl)-5-methoxyindol
3-(2-Hydroxyethyl)-5-methoxyindole
Formel:
C₁₁H₁₃NO₂ Molecular Weight: 191,23 g/mol |
MDL:
MFCD00047196 CAS-Nummer: 712-09-4 EINECS: 211-919-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...