4-Nitro-1H-indazol

Lieferant: BIONET RESEARCH

4P-009.500 4P-009.1 4P-009.5 4P-009.10
BIOT4P-009.500EA 0 EUR
BIOT4P-009.500 BIOT4P-009.1 BIOT4P-009.5 BIOT4P-009.10
4-Nitro-1H-indazol
4-Nitro-1H-indazole
Formel: C₇H₅N₃O₂
Molecular Weight: 163.14 g/mol
MDL: MFCD00022784
CAS-Nummer: 2942-40-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR