2-Amino-6-brombenzothiazol

Lieferant: BIONET RESEARCH

4X-0349.5 4X-0349.500 4X-0349.10 4X-0349.1
BIOT4X-0349.5EA 0 EUR
BIOT4X-0349.5 BIOT4X-0349.500 BIOT4X-0349.10 BIOT4X-0349.1
2-Amino-6-brombenzothiazol
2-Amino-6-bromobenzothiazole
Formel: C₇H₅BrN₂S
Molecular Weight: 229.1 g/mol
MDL: MFCD00152229
CAS-Nummer: 15864-32-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR