4-Brombenzoylacetonitril

Lieferant: BIONET RESEARCH

Synonyme: 3-(4-Bromphenyl)-3-ketopropionsäurenitril, 3-(4-Bromphenyl)-3-oxopropionsäurenitril

4Y-0832.500 4Y-0832.5 4Y-0832.10 4Y-0832.1
BIOT4Y-0832.500EA 0 EUR
BIOT4Y-0832.500 BIOT4Y-0832.5 BIOT4Y-0832.10 BIOT4Y-0832.1
4-Brombenzoylacetonitril
4-Brombenzoylacetonitril
Formel: C₉H₆BrNO
Molecular Weight: 224.06 g/mol
MDL: MFCD00996063
CAS-Nummer: 4592-94-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR