5-(4-Chlorphenyl)-1H-tetrazol
Lieferant: BIONET RESEARCH
Synonyme:
5-(4-Chlorophenyl)-2H-1,2,3,4-tetrazole
6F-900.10
6F-900.500
6F-900.1
6F-900.5
BIOT6F-900.10EA
0
EUR
BIOT6F-900.10
BIOT6F-900.500
BIOT6F-900.1
BIOT6F-900.5
5-(4-Chlorphenyl)-1H-tetrazol
5-(4-Chlorphenyl)-1H-tetrazol
Formel:
C₇H₅ClN₄ Molecular Weight: 180.6 g/mol |
MDL:
MFCD00040952 CAS-Nummer: 16687-61-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...