6-Chloruracil
Lieferant: BIONET RESEARCH
Synonyme:
6-Chlor-2,4-dihydroxypyrimidin, 4-Chlor-2,6-dihydroxypyrimidin, 4-Chloruracil, 6-Chlor-2,4(1H,3H)-pyrimidindion
7K-912.500
7K-912.1
BIOT7K-912.500EA
0
EUR
BIOT7K-912.500
BIOT7K-912.1
6-Chloruracil
6-Chloruracil
Formel:
C₄H₃ClN₂O₂ Molecular Weight: 146.53 g/mol |
MDL:
MFCD00014595 CAS-Nummer: 4270-27-3 EINECS: 224-258-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...