1-(2-Fluor-4-nitrophenyl)piperazin
Lieferant: BIONET RESEARCH
7R-1005.500
7R-1005.1
7R-1005.5
7R-1005.10
BIOT7R-1005.500EA
0
EUR
BIOT7R-1005.500
BIOT7R-1005.1
BIOT7R-1005.5
BIOT7R-1005.10
1-(2-Fluor-4-nitrophenyl)piperazin
1-(2-Fluor-4-nitrophenyl)piperazin
Formel:
C₁₀H₁₂FN₃O₂ |
MDL:
MFCD01572463 CAS-Nummer: 154590-33-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...