2-Amino-5-chlorpyrimidin

Lieferant: BIONET RESEARCH

Synonyme: 5-Chlorpyrimidin-2-ylamin

8P-009.500 8P-009.1 8P-009.5 8P-009.10
BIOT8P-009.500EA 0 EUR
BIOT8P-009.500 BIOT8P-009.1 BIOT8P-009.5 BIOT8P-009.10
2-Amino-5-chlorpyrimidin
2-Amino-5-chloropyrimidine
Formel: C₄H₄ClN₃
Molecular Weight: 129.55 g/mol
MDL: MFCD00014604
CAS-Nummer: 5428-89-7
EINECS: 226-581-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR