6-Chlor-4-nitro-o-toluidin

Lieferant: BIONET RESEARCH
Gefahr

Synonyme: 2-Chlor-6-methyl-4-nitroanilin, 2-Amino-3-chlor-5-nitrotoluol, 6-Chlor-2-methyl-4-nitroanilin

9T-0036.500 9T-0036.1 9T-0036.5 9T-0036.10
BIOT9T-0036.500EA 0 EUR
BIOT9T-0036.500 BIOT9T-0036.1 BIOT9T-0036.5 BIOT9T-0036.10
6-Chlor-4-nitro-o-toluidin
6-Chlor-4-nitro-o-toluidin
Formel: C₇H₇ClN₂O₂
Molecular Weight: 186.6
Schmelzpunkt: 172,5
Lagertemperatur: Raumtemperatur
MDL: MFCD01313703
CAS-Nummer: 69951-02-6
UN: 2237
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR