2-Brom-6-methyl-4-nitroanilin

Lieferant: BIONET RESEARCH

Synonyme: 2-Amino-3-brom-5-nitrotoluol, 6-Brom-4-nitro-o-toluidin

9T-0037.500 9T-0037.1 9T-0037.5 9T-0037.10
BIOT9T-0037.500EA 0 EUR
BIOT9T-0037.500 BIOT9T-0037.1 BIOT9T-0037.5 BIOT9T-0037.10
2-Brom-6-methyl-4-nitroanilin
2-Brom-6-methyl-4-nitroanilin
Formel: C₇H₇BrN₂O₂
Molecular Weight: 231,05 g/mol
MDL: MFCD00053089
CAS-Nummer: 102170-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR