Tricin, Calbiochem®

Lieferant: Merck Millipore Calbiochem

Synonyme: N-(Tris(hydroxymethyl)methyl)glycin

39468-1 39468-100
39468-1EA 724 EUR
39468-1 39468-100
Tricin, Calbiochem®
Tricin
Formel: (HOCH₂)₃CNHCH₂CO₂H
Molecular Weight: 179,17 g/mol
Schmelzpunkt: 187 °C
Dichte: 1,523 g/cm³ (19.85 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004277
CAS-Nummer: 5704-04-1
EINECS: 227-193-6
Merck Index: 13,09724

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR