Propidiumiodid, Sigma-Aldrich®

Lieferant: Merck Millipore Calbiochem
Achtung

Synonyme: 3,8-Diamino-5-(3-(diethylmethylammonio)propyl)-6-phenylphenanthridiniumdiiodid

537059-250EA 477 EUR
537059-250 537059-100 537059-50 537059-1
Propidiumiodid, Sigma-Aldrich®
Propidiumiodid
Formel: C₂₇H₃₄I₂N₄
Molecular Weight: 668,4 g/mol
MDL: MFCD00011921
CAS-Nummer: 25535-16-4
EINECS: 247-081-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR