Anilin-[Ring-D5] (98% D)

Lieferant: LGC Standards PROMOCHEM

Synonyme: Aniline-[ring-²H₅]

CIL-DLM-862-1 CIL-DLM-862-5
PROOCIL-DLM-862-1EA 235 EUR
PROOCIL-DLM-862-1 PROOCIL-DLM-862-5
Anilin-[Ring-D5] (98% D)
Anilin-[Ring-D5]
Formel: C₆H₂D₅N
Molecular Weight: 98,09 g/mol
Schmelzpunkt: –6,3 °C
Dichte: 1,077 g/cm³ (20 °C)
CAS-Nummer: 4165-61-1
EINECS: 224-015-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR